Stampa…

4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-(Hydroxymethyl)-4-methylcyclohexane

M1412-25G M1412-5G
TCIAM1412-25GEA 315 EUR
TCIAM1412-25G TCIAM1412-5G
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture)
Formula: C₈H₁₆O
Peso molecolare: 128,22 g/mol
MDL Number: MFCD00152628
CAS Number: 34885-03-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR