Stampa…

Minoxidil ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten

M1389-5G M1389-1G
TCIAM1389-5GEA 92 EUR
TCIAM1389-5G TCIAM1389-1G
Minoxidil ≥98.0% (by HPLC, titration analysis)
Minoxidil
Formula: C₉H₁₅N₅O
MDL Number: MFCD00063409
CAS Number: 38304-91-5
EINECS: 253-874-2
Merck Index: 14,06203
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e9a2532ee26e29423199440f1aaba2fc6e2ec57&vid=6e9a2532ee26e29423199440f1aaba2fc6e2ec57&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR