Stampa…

Ethyl (4-methoxybenzoyl)acetate ≥97.0% (by GC)

Fornitore: TCI

M1380-25G M1380-5G
TCIAM1380-25GEA 116 EUR
TCIAM1380-25G TCIAM1380-5G
Ethyl (4-methoxybenzoyl)acetate ≥97.0% (by GC)
Ethyl (4-methoxybenzoyl)acetate
Formula: C₁₂H₁₄O₄
Punto di ebollizione: 170°C
Flash Pt: 115°C
MDL Number: MFCD00449636
CAS Number: 2881-83-6
EINECS: 220-733-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eec412e1f6df24105094a2a80c4733858e64b0c&vid=6eec412e1f6df24105094a2a80c4733858e64b0c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR