Stampa…

N-Methyl-D-aspartic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid

M1360-100MG M1360-1G
TCIAM1360-100MGEA 43 EUR
TCIAM1360-100MG TCIAM1360-1G
N-Methyl-D-aspartic acid ≥98.0% (by titrimetric analysis)
N-Methyl-D-aspartic acid
Formula: C₅H₉NO₄
Peso molecolare: 147,13 g/mol
MDL Number: MFCD00004226
CAS Number: 6384-92-5
Merck Index: 14,06662
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab55287b926d779e150b1d453a724da4464a7ade&vid=ab55287b926d779e150b1d453a724da4464a7ade&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -14.0 to -17.0 deg (C=2, H2O)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR