Stampa…

2-Methyl-2-adamantanol ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Hydroxy-2-methyladamantane, 2-Methyladamantan-2-ol

M1334-25G M1334-5G
TCIAM1334-25GEA 135 EUR
TCIAM1334-25G TCIAM1334-5G
2-Methyl-2-adamantanol ≥99.0% (by GC)
2-Methyl-2-adamantanol
Formula: C₁₁H₁₈O
Peso molecolare: 166,26 g/mol
MDL Number: MFCD00074811
CAS Number: 702-98-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 215.0 to 219.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR