5-Methylfurfurylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
M1311-25G
M1311-5G
TCIAM1311-25GEA
255
EUR
TCIAM1311-25G
TCIAM1311-5G
5-Methylfurfurylamine ≥98.0% (by GC, titration analysis)
5-Methylfurfurylamine
Formula:
C₆H₉NO Punto di ebollizione: 55°C Flash Pt: 57°C |
MDL Number:
MFCD00143471 CAS Number: 14003-16-8 UN: 2733 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...