Stampa…

Ethyl-6-methylsalicylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-Hydroxy-6-methylbenzoate, 2-Hydroxy-6-methylbenzoic Acid Ethyl Ester, 6-Methylsalicylic Acid Ethyl Ester

M1283-25G M1283-5G
TCIAM1283-25GEA 345 EUR
TCIAM1283-25G TCIAM1283-5G
Ethyl-6-methylsalicylate ≥98.0% (by GC)
Ethyl-6-methylsalicylate
Formula: C₁₀H₁₂O₃
Peso molecolare: 180,20 g/mol
MDL Number: MFCD01194299
CAS Number: 6555-40-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR