Stampa…

6-Methyl-2-pyridinemethanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxymethyl-6-methylpyridine, 6-Hydroxymethyl-2-picoline

M1282-25G M1282-5G
TCIAM1282-25GEA 440 EUR
TCIAM1282-25G TCIAM1282-5G
6-Methyl-2-pyridinemethanol ≥98.0% (by GC)
6-Methyl-2-pyridinemethanol
Formula: C₇H₉NO
Peso molecolare: 123,16 g/mol
MDL Number: MFCD00023520
CAS Number: 1122-71-0
EINECS: 214-358-2

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR