Stampa…

5-Methoxy-1H-indole-3-carbaldehyde ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

M1268-1G M1268-5G
TCIAM1268-1GEA 52 EUR
TCIAM1268-1G TCIAM1268-5G
5-Methoxy-1H-indole-3-carbaldehyde ≥95.0% (by titrimetric analysis)
5-Methoxy-1H-indole-3-carbaldehyde
Formula: C10H9NO2
Peso molecolare: 175,19 g/mol
MDL Number: MFCD00005623
CAS Number: 10601-19-1
EINECS: 234-220-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Oxime Formation) min. 95.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR