Stampa…

(R)-(+)-Methylsuccinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M1226-25G M1226-5G
TCIAM1226-25GEA 175 EUR
TCIAM1226-25G TCIAM1226-5G
(R)-(+)-Methylsuccinic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-Methylsuccinic acid
Formula: C₅H₈O₄
MDL Number: MFCD00209602
CAS Number: 3641-51-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 97.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR