Stampa…

Sodium-3-mercapto-1-propanesulphonate ≥85.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium 3-mercaptopropanesulphonate, 3-Mercapto-1-propanesulphonic acid Sodium Salt

M1189-25G M1189-500G M1189-100G
TCIAM1189-25GEA 29 EUR
TCIAM1189-25G TCIAM1189-500G TCIAM1189-100G
Sodium-3-mercapto-1-propanesulphonate ≥85.0% (by titrimetric analysis)
Sodium-3-mercapto-1-propanesulphonate
Formula: C₃H₈O₃S₂
Peso molecolare: 178,20 g/mol
MDL Number: MFCD00043148
CAS Number: 17636-10-1
EINECS: 241-620-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90be71ceae0b387f6556df9f028b281f92bc26a6&vid=90be71ceae0b387f6556df9f028b281f92bc26a6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR