Stampa…

N-Methyl-N-(2-pyridyl)formamide ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-(N-Formyl-N-methylamino)pyridine, Meyer's Reagent

M1187-1G M1187-5G
TCIAM1187-1GEA 47 EUR
TCIAM1187-1G TCIAM1187-5G
N-Methyl-N-(2-pyridyl)formamide ≥98.0% (by GC)
N-Methyl-N-(2-pyridyl)formamide
Formula: C₇H₈N₂O
Peso molecolare: 136,15 g/mol
Flash Pt: 109°C
MDL Number: MFCD00006260
CAS Number: 67242-59-5
EINECS: 266-616-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR