Stampa…

4-Methoxy-3-nitrobiphenyl ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Nitro-4-phenylanisole

M1166-25G M1166-5G
TCIAM1166-25GEA 141 EUR
TCIAM1166-25G TCIAM1166-5G
4-Methoxy-3-nitrobiphenyl ≥99.0% (by GC)
4-Methoxy-3-nitrobiphenyl
Formula: C₁₃H₁₁NO₃
Peso molecolare: 229,24 g/mol
MDL Number: MFCD00191554
CAS Number: 15854-73-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 88.0 to 90.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR