Stampa…

2,2'-Methylenebis(4-methylphenol) ≥90.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-Methylenebis(p-cresol), 2,2'-Dihydroxy-5,5'-dimethyldiphenylmethane, Bis(2-hydroxy-5-methylphenyl)methane

M1134-25G M1134-5G
TCIAM1134-25GEA 124 EUR
TCIAM1134-25G TCIAM1134-5G
2,2'-Methylenebis(4-methylphenol) ≥90.0% (by GC)
2,2'-Methylenebis(4-methylphenol)
Formula: C₁₅H₁₆O₂
Peso molecolare: 228,29 g/mol
MDL Number: MFCD00155176
CAS Number: 3236-63-3
EINECS: 221-794-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7f4c737212d22c87a5c48025d4f3761096ed1b6&vid=c7f4c737212d22c87a5c48025d4f3761096ed1b6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Melting point 123.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR