tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(N-Boc-N-methylamino)pyrrolidine, 3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine
M1096-25G
M1096-5G
TCIAM1096-25GEA
704
EUR
TCIAM1096-25G
TCIAM1096-5G
tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥96.0% (by GC)
tert-Butyl methyl(pyrrolidin-3-yl)carbamate
Formula:
C₁₀H₂₀N₂O₂ Peso molecolare: 200,28 g/mol Punto di ebollizione: 96°C |
MDL Number:
MFCD00191665 CAS Number: 172478-00-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...