Stampa…

Methylsulphamic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

M1083-25G M1083-5G
TCIAM1083-25GEA 615 EUR
TCIAM1083-25G TCIAM1083-5G
Methylsulphamic acid ≥98.0% (by titrimetric analysis)
Methylsulphamic acid
Formula: CH₅NO₃S
MDL Number: MFCD00010870
CAS Number: 4112-03-2
EINECS: 223-895-1
UN: 1759
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR