Stampa…

2-Ethylhexyl-4-methoxycinnamate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Methoxycinnamic Acid 2-Ethylhexyl Ester, 4-Methoxycinnamic Acid Octyl Ester, Octinoxate, Octyl 4-Methoxycinnamate, 2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

M1082-25ML M1082-500ML M1082-100ML
TCIAM1082-25MLEA 19 EUR
TCIAM1082-25ML TCIAM1082-500ML TCIAM1082-100ML
2-Ethylhexyl-4-methoxycinnamate ≥97.0% (by GC)
2-Ethylhexyl-4-methoxycinnamate
Formula: C₁₈H₂₆O₃
Peso molecolare: 290,40 g/mol
Flash Pt: 193°C
MDL Number: MFCD00072582
CAS Number: 5466-77-3
EINECS: 226-775-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465ef7f7e693296de54d81654b6069a1e66cc88c&vid=465ef7f7e693296de54d81654b6069a1e66cc88c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR