Stampa…

Monomyristin ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Monomyristoyl Glycerol, Glycerol alpha-Monomyristate, 2,3-Dihydroxypropyl tetradecanoate

M1073-1G M1073-5G
TCIAM1073-1GEA 69 EUR
TCIAM1073-1G TCIAM1073-5G
Monomyristin ≥97.0% (by GC)
Monomyristin
Formula: C₁₇H₃₄O₄
Peso molecolare: 302,46 g/mol
MDL Number: MFCD00046760
CAS Number: 589-68-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 69.0 to 72.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR