Stampa…

2-Ethylhexyl-3-mercaptopropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 2-Ethylhexyl 3-mercaptopropanoate, 2-Octyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Octyl Ester

M1067-25ML M1067-500ML
TCIAM1067-25MLEA 17 EUR
TCIAM1067-25ML TCIAM1067-500ML
2-Ethylhexyl-3-mercaptopropionate ≥98.0% (by GC)
2-Ethylhexyl-3-mercaptopropionate
Formula: C₁₁H₂₂O₂S
Peso molecolare: 218,36 g/mol
Flash Pt: 125°C
MDL Number: MFCD00087933
CAS Number: 50448-95-8
EINECS: 256-589-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR