Stampa…

N-Methyl-3-chloropropylamine hydrochloride ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 3-(Methylamino)propyl Chloride Hydrochloride, 3-Chloro-N-methylpropan-1-amine hydrochloride

M1048-1G M1048-5G
TCIAM1048-1GEA 79 EUR
TCIAM1048-1G TCIAM1048-5G
N-Methyl-3-chloropropylamine hydrochloride ≥99.0% (by titrimetric analysis)
N-Methyl-3-chloropropylamine hydrochloride
Formula: C₄H₁₀ClN
Peso molecolare: 144,04 g/mol
MDL Number: MFCD00144935
CAS Number: 97145-88-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 103.0 to 106.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR