Benzyl methyl malonate ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Malonic Acid Benzyl Methyl Ester
M0974-25G
M0974-5G
TCIAM0974-25GEA
142
EUR
TCIAM0974-25G
TCIAM0974-5G
Benzyl methyl malonate ≥96.0% (by GC)
Benzyl methyl malonate
Formula:
C₁₁H₁₂O₄ Peso molecolare: 208,21 g/mol |
MDL Number:
MFCD00008461 CAS Number: 52267-39-7 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...