(S)-(+)-3-Methyl-1-pentanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
M0963-1ML
M0963-5ML
TCIAM0963-1MLEA
106
EUR
TCIAM0963-1ML
TCIAM0963-5ML
(S)-(+)-3-Methyl-1-pentanol ≥98.0% (by GC)
(S)-(+)-3-Methyl-1-pentanol
Formula:
C₆H₁₄O Peso molecolare: 102,18 g/mol Punto di ebollizione: 152°C |
MDL Number:
MFCD00135167 CAS Number: 42072-39-9 UN: 1987 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +8.5 to +8.8 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...