Stampa…

4-Morpholinylcarbonyl chloride ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Morpholinocarbonyl Chloride, 4-Chlorocarbonylmorpholine, Morpholine-4-carbonyl chloride, 4-Morpholine-carbonylchloride

M0935-25G M0935-100G
TCIAM0935-25GEA 89 EUR
TCIAM0935-25G TCIAM0935-100G
4-Morpholinylcarbonyl chloride ≥97.0% (by GC, titration analysis)
4-Morpholinylcarbonyl chloride
Formula: C₅H₈ClNO₂
Peso molecolare: 149,57 g/mol
Punto di ebollizione: 138°C
MDL Number: MFCD00037053
CAS Number: 15159-40-7
EINECS: 239-213-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6329857bdeb2843a74db3e8ef81253ebdc1531f6&vid=6329857bdeb2843a74db3e8ef81253ebdc1531f6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR