Stampa…

N-Methoxy-N-methylacetamide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M0919-10G M0919-25G
TCIAM0919-10GEA 69 EUR
TCIAM0919-10G TCIAM0919-25G
N-Methoxy-N-methylacetamide ≥98.0% (by GC)
N-Methoxy-N-methylacetamide
Formula: C₄H₉NO₂
Peso molecolare: 103,12 g/mol
Punto di ebollizione: 94°C
Flash Pt: 49°C
MDL Number: MFCD00060098
CAS Number: 78191-00-1
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR