Methacroylcholine chloride ∼80% in water stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(Methacryloyloxy)-N,N,N-trimethylethanaminium chloride, (Methacryloyloxyethyl)trimethyl ammonium chloride, 2-(Methacryloyloxy)ethyltrimethylammonium chloride, Trimethyl-2-methacroyloxyethylammonium Chloride
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₉H₁₈ClNO₂ Peso molecolare: 207,70 g/mol |
MDL Number:
MFCD00060097 CAS Number: 5039-78-1 EINECS: 225-733-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Bromination Method) | 79.0 to 87.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...