2-Methyl-5-isopropylaniline ≥93.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
5-Isopropyl-2-methylaniline, 2-Amino-p-cymene, 5-Isopropyl-o-toluidine
M0849-1G
M0849-5G
TCIAM0849-1GEA
102
EUR
TCIAM0849-1G
TCIAM0849-5G
2-Methyl-5-isopropylaniline ≥93.0% (by GC)
2-Methyl-5-isopropylaniline
Formula:
C₁₀H₁₅N Peso molecolare: 149,24 g/mol |
MDL Number:
MFCD00044503 CAS Number: 2051-53-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...