2-Methyl-3-butenenitrile ≥75.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₅H₇N Punto di ebollizione: 124°C Flash Pt: 15°C |
MDL Number:
MFCD00042634 CAS Number: 16529-56-9 EINECS: 240-596-1 UN: 3273 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 75.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...