Stampa…

Dimethyl methoxymalonate ≥95.0% (by GC)

Fornitore: TCI
Attenzione

M0836-25G M0836-5G
TCIAM0836-25GEA 345 EUR
TCIAM0836-25G TCIAM0836-5G
Dimethyl methoxymalonate ≥95.0% (by GC)
Dimethyl methoxymalonate
Formula: C₆H₁₀O₅
Punto di ebollizione: 110°C
Flash Pt: 107°C
MDL Number: MFCD00009847
CAS Number: 5018-30-4
EINECS: 225-697-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1684c8c632e61e4ca68ccf0bd1f25a778b4d1d88&vid=1684c8c632e61e4ca68ccf0bd1f25a778b4d1d88&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR