Stampa…

Methyl-5-methoxy-3-oxovalerate ≥96.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate, 3-Keto-5-methoxyvaleric Acid Methyl Ester, Methyl 5-methoxy-3-oxopentanoate

M0812-1G M0812-5G
TCIAM0812-1GEA 128 EUR
TCIAM0812-1G TCIAM0812-5G
Methyl-5-methoxy-3-oxovalerate ≥96.0% (by GC)
Methyl-5-methoxy-3-oxovalerate
Formula: C₇H₁₂O₄
Peso molecolare: 160,17 g/mol
Punto di ebollizione: 69°C
Flash Pt: 104°C
MDL Number: MFCD00042840
CAS Number: 62462-05-9
EINECS: 263-553-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR