3-Methylthiopropylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₄H₁₁NS Punto di ebollizione: 170°C Flash Pt: 62°C |
MDL Number:
MFCD00041898 CAS Number: 4104-45-4 EINECS: 223-875-2 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...