Tetrahydrofurfuryl methacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(Tetrahydrofuran-2-yl)methyl methacrylate, Methacrylic Acid Tetrahydrofurfuryl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
M0745-25ML
M0745-500ML
TCIAM0745-25MLEA
16
EUR
TCIAM0745-25ML
TCIAM0745-500ML
Tetrahydrofurfuryl methacrylate ≥98.0% (by GC) stabilizzato
Tetrahydrofurfuryl methacrylate
Formula:
C₉H₁₄O₃ Peso molecolare: 170,21 g/mol Punto di ebollizione: 85°C Flash Pt: 96°C |
MDL Number:
MFCD00014515 CAS Number: 2455-24-5 EINECS: 219-529-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...