Stampa…

Butyltin hydroxide oxide

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-butyltinhydroxideoxide, Butylhydroxyoxostannane

M0744-25G M0744-500G
TCIAM0744-25GEA 18 EUR
TCIAM0744-25G TCIAM0744-500G
Butyltin hydroxide oxide
Butyltin hydroxide oxide
Formula: C₄H₁₀O₂Sn
Peso molecolare: 208,83 g/mol
MDL Number: MFCD00013927
CAS Number: 2273-43-0
EINECS: 218-880-1
UN: 3146
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Drying loss max. 3.0 %
Ignition residue (as SnO2) 71.0 to 73.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR