Stampa…

N-Methylisatoic anhydride ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

M0743-25G M0743-5G
TCIAM0743-25GEA 330 EUR
TCIAM0743-25G TCIAM0743-5G
N-Methylisatoic anhydride ≥98.0% (by GC)
N-Methylisatoic anhydride
Formula: C₉H₇NO₃
Peso molecolare: 177,16 g/mol
MDL Number: MFCD00006815
CAS Number: 10328-92-4
EINECS: 233-714-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13df4a45ba3eb0f620e54cabd0d40099ca6e6b7a&vid=13df4a45ba3eb0f620e54cabd0d40099ca6e6b7a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR