Stampa…

Vinyl methacrylate ≥98.0% (by GC) stabilizzato

Fornitore: TCI
Pericolo


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0720-25ML M0720-500ML M0720-100ML
TCIAM0720-25MLEA 29 EUR
TCIAM0720-25ML TCIAM0720-500ML TCIAM0720-100ML
Vinyl methacrylate ≥98.0% (by GC) stabilizzato
Vinyl methacrylate
Formula: C₆H₈O₂
Punto di ebollizione: 112°C
Flash Pt: 16°C
MDL Number: MFCD00059329
CAS Number: 4245-37-8
EINECS: 224-205-1
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=547cd9c4eeb3ce429e81819ee278711589bf62df&vid=547cd9c4eeb3ce429e81819ee278711589bf62df&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR