Stampa…

(2-Methoxyphenyl)hydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Methoxyphenylhydrazine hydrochloride, 2-Hydrazinoanisole hydrochloride

M0617-25G M0617-5G
TCIAM0617-25GEA 582 EUR
TCIAM0617-25G TCIAM0617-5G
(2-Methoxyphenyl)hydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)
(2-Methoxyphenyl)hydrazine hydrochloride
Formula: C₇H₁₁ClN₂O
Peso molecolare: 174,6280 g/mol
MDL Number: MFCD00035456
CAS Number: 6971-45-5
EINECS: 230-199-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR