Ethyl hydrogen maleate ≥95.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₈O₄ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00063175 CAS Number: 3990-03-2 EINECS: 223-635-7 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...