Cyclohexyl methacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methacrylic Acid Cyclohexyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
M0594-25ML
M0594-500ML
TCIAM0594-25MLEA
17
EUR
TCIAM0594-25ML
TCIAM0594-500ML
Cyclohexyl methacrylate ≥98.0% (by GC) stabilizzato
Cyclohexyl methacrylate
Formula:
C₁₀H₁₆O₂ Peso molecolare: 168,24 g/mol Punto di ebollizione: 72°C Flash Pt: 78°C |
MDL Number:
MFCD00014292 CAS Number: 101-43-9 EINECS: 202-943-5 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Appearance | Colorless to Almost colorless clear liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...