Stampa…

2-(Diethylamino)ethyl methacrylate ≥98.5% (by GC) stabilizzato

Fornitore: TCI
Attenzione

Synonyms: Methacrylic Acid 2-(Diethylamino)ethyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0562-25ML M0562-500ML
TCIAM0562-25MLEA 16 EUR
TCIAM0562-25ML TCIAM0562-500ML
2-(Diethylamino)ethyl methacrylate ≥98.5% (by GC) stabilizzato
2-(Diethylamino)ethyl methacrylate
Formula: C₁₀H₁₉NO₂
Peso molecolare: 185.27 g/mol
Flash Pt: 94°C
MDL Number: MFCD00038314
CAS Number: 105-16-8
EINECS: 203-275-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR