Stampa…

Dibutyl malonate ≥99.0% (by GC)

Fornitore: TCI

Synonyms: Malonic Acid Dibutyl Ester

M0540-25G M0540-500G
TCIAM0540-25GEA 52 EUR
TCIAM0540-25G TCIAM0540-500G
Dibutyl malonate ≥99.0% (by GC)
Dibutyl malonate
Formula: C₁₁H₂₀O₄
Punto di ebollizione: 251°C
MDL Number: MFCD00053795
CAS Number: 1190-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf40f1d477c09fc6ece9466c2c923d225249daff&vid=cf40f1d477c09fc6ece9466c2c923d225249daff&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR