Stampa…

3-Methyl-1H-pyrazol-5(4H)-one ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-methyl-4,5-dihydro-1H-pyrazol-5-one

M0491-25G M0491-500G
TCIAM0491-25GEA 29 EUR
TCIAM0491-25G TCIAM0491-500G
3-Methyl-1H-pyrazol-5(4H)-one ≥98.0% (by titrimetric analysis)
3-Methyl-1H-pyrazol-5(4H)-one
Formula: C4H6N2O
Peso molecolare: 98,11 g/mol
MDL Number: MFCD00020699
CAS Number: 108-26-9
EINECS: 203-565-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 220.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR