Stampa…

Myristic anhydride ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Myristic anhydride

M0484-25G M0484-250G
TCIAM0484-25GEA 77 EUR
TCIAM0484-25G TCIAM0484-250G
Myristic anhydride ≥95.0% (by titrimetric analysis)
Myristic anhydride
Formula: C₂₈H₅₄O₃
Peso molecolare: 438,74 g/mol
MDL Number: MFCD00056151
CAS Number: 626-29-9
EINECS: 210-941-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 95.0 %
Melting point 53.0 to 55.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR