Butyl myristate ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
M0477-25ML
M0477-500ML
TCIAM0477-25MLEA
41
EUR
TCIAM0477-25ML
TCIAM0477-500ML
Butyl myristate ≥97.0% (by GC)
Butyl myristate
Formula:
C₁₈H₃₆O₂ Punto di ebollizione: 195°C Flash Pt: 166°C |
MDL Number:
MFCD00015077 CAS Number: 110-36-1 EINECS: 203-759-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...