Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)
Formula:
C₁₀H₁₄MoO₆ Peso molecolare: 326.18 g/mol |
MDL Number:
MFCD00011506 CAS Number: 17524-05-9 EINECS: 241-522-0 |
|
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Solubility in hot EtOH | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...