Isobutyl mercaptan ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₄H₁₀S Punto di ebollizione: 88°C Flash Pt: -10°C |
MDL Number:
MFCD00004882 CAS Number: 513-44-0 EINECS: 208-162-6 UN: 2347 ADR: 3,II Merck Index: 14,05147 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...