1-(4-Nitrophenyl)-3-methyl-5-pyrazolone ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Synonyms:
5-Methyl-2-(4-nitrophenyl)-2,4-dihydro-3H-pyrazol-3-one, 3-Methyl-1-(4-nitrophenyl)-5-pyrazolone
Formula:
C₁₀H₉N₃O₃ |
MDL Number:
MFCD00020751 CAS Number: 6402-09-1 EINECS: 229-018-9 |
Risultati del test di specificazione
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...