Stampa…

5-Methylcytosine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 5-Methylcytosinehydrochloride

M0204-1G M0204-100MG M0204-5G
TCIAM0204-1GEA 131 EUR
TCIAM0204-1G TCIAM0204-100MG TCIAM0204-5G
5-Methylcytosine hydrochloride ≥98.0% (by HPLC, total nitrogen)
5-Methylcytosine hydrochloride
Formula: C₅H₇N₃O
Peso molecolare: 161,59 g/mol
MDL Number: MFCD00035313
CAS Number: 58366-64-6
EINECS: 261-223-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR