Stampa…

Isobutyl methacrylate polymer

Fornitore: TCI

Synonyms: Methacrylic Acid Isobutyl Ester Polymer, Poly(isobutyl methacrylate)

M0086-25G M0086-500G
TCIAM0086-25GEA 16 EUR
TCIAM0086-25G TCIAM0086-500G
Isobutyl methacrylate polymer
Isobutyl methacrylate polymer
Formula: (C₈H₁₄O₂)n
MDL Number: MFCD00008931
CAS Number: 9011-15-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Viscosity 100.0 to 160.0 mPa-s (40 wt%, Toluene, 25 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR