Methacholine bromide ≥99.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
O-Acetyl-beta-methylcholine Bromide, Acetyl-β-methylcholine bromide, Mecholyl Bromide
Formula:
C₈H₁₈BrNO₂ Peso molecolare: 240.14 g/mol |
MDL Number:
MFCD00011816 CAS Number: 333-31-3 EINECS: 206-372-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Purity (Argentmetric Titration) | min. 99.0 % |
Melting point | 149.0 to 153.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...