Stampa…

6-Mercaptopurine monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

M0063-1G M0063-5G
TCIAM0063-1GEA 16 EUR
TCIAM0063-1G TCIAM0063-5G
6-Mercaptopurine monohydrate ≥98.0% (by HPLC, titration analysis)
6-Mercaptopurine monohydrate
Formula: C₅H₄N₄S·H₂O
MDL Number: MFCD03854445
CAS Number: 6112-76-1
Merck Index: 14,05871

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Water 9.9 to 11.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR