Stampa…

DL-Malic acid disodium salt ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Di-Sodium Dl-Malate, DL-Malic acid disodium salt, DL-Apple acid disodium salt, Disodium DL-Malate hydrate

M0026-25G M0026-500G
TCIAM0026-25GEA 16 EUR
TCIAM0026-25G TCIAM0026-500G
DL-Malic acid disodium salt ≥98.0% (by titrimetric analysis)
DL-Malic acid disodium salt
Formula: C₄H₄Na₂O₅
MDL Number: MFCD00012466
CAS Number: 676-46-0
EINECS: 211-627-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR